Learn More
Aviva Systems Biology Corp UCK2 Antibody - N-terminal region (ARP46184_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46184_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UCK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UCK2. Alternate names: TSA903, UK, UMPKApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-59262