Learn More
Medchemexpress LLC UCL 1684 dibromide | 199934-16-2 | 98.1% | 654.44 g/mol | C34H30Br2N4 | 5 MG
SDP
Catalog No. 5000264776
Encompass_Preferred
UCL 1684 dibromide is a potent, non-peptidic, nanomolar inhibitor of small conductance calcium-activated potassium (SK) channels, intended for research use in electrophysiology and ion channel studies.
  • Potent SK channel blocker with nanomolar activity.
  • High purity (98.1%) suitable for biochemical and electrophysiological assays.
  • Molecular weight 654.44 g/mol for formulation calculations.
  • Solid, white to off-white material for convenient handling and weighing.
  • Stable when stored sealed and protected from moisture; follow recommended cold storage for solutions.
Catalog No. 50-002-64776 Supplier Medchemexpress LLC Supplier No. HY1085795MG
UCL 1684 dibromide is a potent, non-peptidic, nanomolar inhibitor of small conductance calcium-activated potassium (SK) channels, intended for research use in electrophysiology and ion channel studies.
  • Potent SK channel blocker with nanomolar activity.
  • High purity (98.1%) suitable for biochemical and electrophysiological assays.
  • Molecular weight 654.44 g/mol for formulation calculations.
  • Solid, white to off-white material for convenient handling and weighing.
  • Stable when stored sealed and protected from moisture; follow recommended cold storage for solutions.