Learn More
TARGETMOL CHEMICALS INC UCL 2077 5MG
SDP
Catalog No. 502581115
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UCL 2077 is a subtype-selective blocker of the epilepsy-associated KCNQ channels and it also is a selective slow-afterhyperpolarization channel blocker,with IC50 of 500 nM in hippocampal neurons in culture having minimal effects on Ca2+ channels action potentials input resistance and the medium afterhyperpolarization. purity: 99%
Catalog No. 50-258-1115 Supplier TARGETMOL CHEMICALS INC Supplier No. T171975MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UCL 2077 is a subtype-selective blocker of the epilepsy-associated KCNQ channels and it also is a selective slow-afterhyperpolarization channel blocker,with IC50 of 500 nM in hippocampal neurons in culture having minimal effects on Ca2+ channels action potentials input resistance and the medium afterhyperpolarization. purity: 99%