Learn More
Medchemexpress LLC UCL-TRO-1938 5MG
SDP
Catalog No. NC2596999
Encompass_Preferred
UCL-TRO-1938 is a potent allosteric activator of PI3Kα with an EC50 value of approximately 60 μM. UCL-TRO-1938 can induce cell proliferation and has cardioprotective and neural regeneration effects.
Catalog No. NC2596999 Supplier Medchemexpress LLC Supplier No. HY1548485MG
UCL-TRO-1938 is a potent allosteric activator of PI3Kα with an EC50 value of approximately 60 μM. UCL-TRO-1938 can induce cell proliferation and has cardioprotective and neural regeneration effects.