Learn More
Cayman Chemical UCLA gp130 2 25mg
SDP
Catalog No. 502910095
Encompass_Preferred
A humanin peptide mimetic; inhibits NMDA-induced LDH release in primary mouse hippocampal neurons at 10 µM, an effect that can be reversed by SC144
Catalog No. 50-291-0095 Supplier Cayman Chemical Supplier No. 3555125 MG
A humanin peptide mimetic; inhibits NMDA-induced LDH release in primary mouse hippocampal neurons at 10 µM, an effect that can be reversed by SC144