Learn More
Medchemexpress LLC UCM-05194 ammonium | 482.35 | 1 MG
SDP
Catalog No. 5000390647
Encompass_Preferred
UCM-05194 ammonium is a lysophosphatidic acid receptor 1 (LPA1) agonist. It induces calcium mobilization in LPA1-expressing RH7777 cells and also induces neurite retraction and migration in LPA1-overexpressing B103 rat neuroblastoma cells. This compound attenuates acetic acid-induced writhing and hind paw mechanical hypersensitivity in mice.
  • Acts as a lysophosphatidic acid receptor 1 (LPA1) agonist
  • Induces calcium mobilization in LPA1-expressing RH7777 cells
  • Induces neurite retraction and migration in LPA1-overexpressing B103 rat neuroblastoma cells
  • Attenuates acetic acid-induced writhing in mice
  • Attenuates hind paw mechanical hypersensitivity in mice
Catalog No. 50-003-90647 Supplier Medchemexpress LLC Supplier No. HY167862A1MG
UCM-05194 ammonium is a lysophosphatidic acid receptor 1 (LPA1) agonist. It induces calcium mobilization in LPA1-expressing RH7777 cells and also induces neurite retraction and migration in LPA1-overexpressing B103 rat neuroblastoma cells. This compound attenuates acetic acid-induced writhing and hind paw mechanical hypersensitivity in mice.
  • Acts as a lysophosphatidic acid receptor 1 (LPA1) agonist
  • Induces calcium mobilization in LPA1-expressing RH7777 cells
  • Induces neurite retraction and migration in LPA1-overexpressing B103 rat neuroblastoma cells
  • Attenuates acetic acid-induced writhing in mice
  • Attenuates hind paw mechanical hypersensitivity in mice