Learn More
Medchemexpress LLC UCM-13369 dihydrochloride | 99.87% | 462.46 | 5 MG
SDP
Catalog No. 5000394441
Encompass_Preferred
UCM-13369 (Compound 4b) is a NPM1 inhibitor. It downregulates the pathway associated with mutant NPM1 C+, and specifically recognizes the C-end DNA-binding domain of NPM1 C+. It induces apoptosis in AML cell lines and primary cells and can be used for leukemia research.
  • Downregulates the pathway associated with mutant NPM1 C+.
  • Specifically recognizes the C-end DNA-binding domain of NPM1 C+.
  • Induces apoptosis in AML cell lines and primary cells.
  • Can be used for leukemia research.
Catalog No. 50-003-94441 Supplier Medchemexpress LLC Supplier No. HY162265A5MG
UCM-13369 (Compound 4b) is a NPM1 inhibitor. It downregulates the pathway associated with mutant NPM1 C+, and specifically recognizes the C-end DNA-binding domain of NPM1 C+. It induces apoptosis in AML cell lines and primary cells and can be used for leukemia research.
  • Downregulates the pathway associated with mutant NPM1 C+.
  • Specifically recognizes the C-end DNA-binding domain of NPM1 C+.
  • Induces apoptosis in AML cell lines and primary cells.
  • Can be used for leukemia research.