Learn More
Medchemexpress LLC UCM-13369 dihydrochloride | 99.9% | 462.46 | 25 MG
SDP
Catalog No. 5000395210
Encompass_Preferred
UCM-13369 (Compound 4b) is a NPM1 inhibitor. It downregulates the pathway associated with mutant NPM1 C+ and specifically recognizes the C-end DNA-binding domain of NPM1 C+. It induces apoptosis in AML cell lines and primary cells and can be used for leukemia research.
  • NPM1 inhibitor
  • Downregulates pathway associated with mutant NPM1 C+
  • Specifically recognizes the C-end DNA-binding domain of NPM1 C+
  • Induces apoptosis in AML cell lines and primary cells
  • Can be used for leukemia research
Catalog No. 50-003-95210 Supplier Medchemexpress LLC Supplier No. 25MGHY162265A
UCM-13369 (Compound 4b) is a NPM1 inhibitor. It downregulates the pathway associated with mutant NPM1 C+ and specifically recognizes the C-end DNA-binding domain of NPM1 C+. It induces apoptosis in AML cell lines and primary cells and can be used for leukemia research.
  • NPM1 inhibitor
  • Downregulates pathway associated with mutant NPM1 C+
  • Specifically recognizes the C-end DNA-binding domain of NPM1 C+
  • Induces apoptosis in AML cell lines and primary cells
  • Can be used for leukemia research