Learn More
Medchemexpress LLC Ucm-13369 (dihydrochloride) | 99.9% | 462.46 g/mol | C25H33Cl2N3O | 10 MG
SDP
Catalog No. 5000288486
Encompass_Preferred
UCM-13369 dihydrochloride is a research-grade small-molecule inhibitor of nucleophosmin 1 (NPM1) used for in vitro studies of apoptosis and AML-related pathways. It is supplied as a solid with confirmed high purity and solubility properties for DMSO stock preparation.
  • Inhibits NPM1-associated pathways and induces apoptosis in AML models.
  • High purity (99.87%) suitable for research applications.
  • Available in multiple pack sizes, including 10 mg.
  • Soluble in DMSO at 200 mg/mL for stock solution preparation.
  • Powder stable at -20°C for long-term storage; solutions require cold storage.
Catalog No. 50-002-88486 Supplier Medchemexpress LLC Supplier No. HY162265A10MG
UCM-13369 dihydrochloride is a research-grade small-molecule inhibitor of nucleophosmin 1 (NPM1) used for in vitro studies of apoptosis and AML-related pathways. It is supplied as a solid with confirmed high purity and solubility properties for DMSO stock preparation.
  • Inhibits NPM1-associated pathways and induces apoptosis in AML models.
  • High purity (99.87%) suitable for research applications.
  • Available in multiple pack sizes, including 10 mg.
  • Soluble in DMSO at 200 mg/mL for stock solution preparation.
  • Powder stable at -20°C for long-term storage; solutions require cold storage.