Learn More
TARGETMOL CHEMICALS INC UCM05 5MG
SDP
Catalog No. 502580999
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UCM05 (G28UCM) (G28UCM) is a potent fatty acid synthase ( FASN ) inhibitor that shows activity against HER2+ breast cancer xenografts and is active in anti-HER2 drug-resistant cell lines [1]. UCM05 is a Filamentous temperature-sensitive protein Z (FtsZ) inhibitor and inhibits the growth of the Gram-positive bacterium B. subtilis with MIC values of 100 uM but lack activity on the Gram-negative bacterium E. coli [2]. purity: 99%
Catalog No. 50-258-0999 Supplier TARGETMOL CHEMICALS INC Supplier No. T218315MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UCM05 (G28UCM) (G28UCM) is a potent fatty acid synthase ( FASN ) inhibitor that shows activity against HER2+ breast cancer xenografts and is active in anti-HER2 drug-resistant cell lines [1]. UCM05 is a Filamentous temperature-sensitive protein Z (FtsZ) inhibitor and inhibits the growth of the Gram-positive bacterium B. subtilis with MIC values of 100 uM but lack activity on the Gram-negative bacterium E. coli [2]. purity: 99%