Learn More
Aviva Systems Biology Corp UCP1 Antibody - C-terminal region (ARP63203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UCP1. It was validated on Western Blot. . Alternate names: SLC25A7, UCPApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-287-910