Learn More
Biomatik Corporation Ucp1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10642
Encompass_Preferred
Ucp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to UCP1. The Ucp1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mitochondrial Brown Fat Uncoupling Protein 1 Protein (1-307Aa). Applications: ELISA, WB, IHC. Target Synonyms: UCP1. Accession Number: P25874. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5096