Learn More
Medchemexpress LLC UCPH-101 | 1118460-77-7 | 98.6% | 422.48 | 10 MG
SDP
Catalog No. 5000346567
Encompass_Preferred
UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM. It functions as an allosteric modulator, inducing sustained inhibition of EAAT1 through an intramonomeric site within the trimerization domain. In vitro studies show that UCPH-101 inhibits EAAT1 anion currents in a concentration-dependent manner, with a KD value of 0.34±0.03 μM. This compound is intended for research use only.
  • Inhibits excitatory amino acid transporter subtype 1 (EAAT1)
  • Functions as an allosteric modulator
  • Demonstrates sustained inhibition of EAAT1 anion currents
  • IC50 of 0.66 μM
  • KD value of 0.34±0.03 μM for EAAT1 anion currents
Catalog No. 50-003-46567 Supplier Medchemexpress LLC Supplier No. HY1091450MG
UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM. It functions as an allosteric modulator, inducing sustained inhibition of EAAT1 through an intramonomeric site within the trimerization domain. In vitro studies show that UCPH-101 inhibits EAAT1 anion currents in a concentration-dependent manner, with a KD value of 0.34±0.03 μM. This compound is intended for research use only.
  • Inhibits excitatory amino acid transporter subtype 1 (EAAT1)
  • Functions as an allosteric modulator
  • Demonstrates sustained inhibition of EAAT1 anion currents
  • IC50 of 0.66 μM
  • KD value of 0.34±0.03 μM for EAAT1 anion currents