Learn More
Medchemexpress LLC UCPH-101 | 1118460-77-7 | 98.6% | 422.48 | 50 MG
SDP
Catalog No. 5000344494
Encompass_Preferred
UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM. It demonstrates cellular effects on HEK293 cells by inhibiting human EAAT1. This product is for research use only.
  • Acts as an EAAT1 inhibitor
  • Demonstrates cellular effects on HEK293 cells
  • Inhibits human EAAT1 anion currents
Catalog No. 50-003-44494 Supplier Medchemexpress LLC Supplier No. HY1091410MG
UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM. It demonstrates cellular effects on HEK293 cells by inhibiting human EAAT1. This product is for research use only.
  • Acts as an EAAT1 inhibitor
  • Demonstrates cellular effects on HEK293 cells
  • Inhibits human EAAT1 anion currents