Learn More
Medchemexpress LLC HY-10914 5mg Medchemexpress, UCPH-101 CAS:1118460-77-7 Purity:>98%
SDP
Catalog No. 501968686
Encompass_Preferred
Medchemexpress, HY-10914 5mg UCPH-101 CAS:1118460-77-7 UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8686 Supplier Medchemexpress LLC Supplier No. HY109145MG
Medchemexpress, HY-10914 5mg UCPH-101 CAS:1118460-77-7 UCPH-101 is an excitatory amino acid transporter subtype 1 (EAAT1) inhibitor with an IC50 of 0.66 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.