Learn More
Abcam UCPH-101, Selective EAAT1 inhibitor, 1MG
Catalog No. 502584626
Encompass_Preferred
MW 422.5 Da, Purity >98%. Selective inhibitor of EAAT1 (IC₅₀ = 0.67 μM). Displays >400-fold selectivity over EAAT2 and EAAT3. Blood-brain barrier permeable analog UCPH-102 also available (ab146404).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4626 Supplier Abcam Supplier No. AB1203091MG
MW 422.5 Da, Purity >98%. Selective inhibitor of EAAT1 (IC₅₀ = 0.67 μM). Displays >400-fold selectivity over EAAT2 and EAAT3. Blood-brain barrier permeable analog UCPH-102 also available (ab146404).
The product is subject to the following: Abcam Restricted Use Statement