Learn More
Aviva Systems Biology Corp UCRC Antibody - middle region (ARP44448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UCRC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UCRC. Alternate names: HSPC051, HSPC119, HSPC151, QCR9, UCRC, UCCR7.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58813