Learn More
UCT LLC Enviro-Clean 521 2000mg Activated Carbon, 6 mL
SDP
Catalog No. NC1920247
Encompass
30 x Carbon Extraction Cartridges, Enviro-Clean 521 2000mg Activated Carbon, 6 mL
Catalog No. NC1920247 Supplier UCT LLC Supplier No. EU52112M6
30 x Carbon Extraction Cartridges, Enviro-Clean 521 2000mg Activated Carbon, 6 mL