Learn More
Medchemexpress LLC HY-18253 5mg Medchemexpress, Udenafil CAS:268203-93-6 Purity:>98%
SDP
Catalog No. 502082028
Encompass_Preferred
Medchemexpress, HY-18253 5mg Udenafil CAS:268203-93-6 Udenafil (DA8159) is a potent, selective and orally active phosphodiesterase type 5 (PDE5) inhibitor. Udenafil also inhibits cGMP hydrolysis and can be used for erectile dysfunction research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2028 Supplier Medchemexpress LLC Supplier No. HY182535MG
Medchemexpress, HY-18253 5mg Udenafil CAS:268203-93-6 Udenafil (DA8159) is a potent, selective and orally active phosphodiesterase type 5 (PDE5) inhibitor. Udenafil also inhibits cGMP hydrolysis and can be used for erectile dysfunction research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.