Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Scientific UDK129 Distillation Unit
SDP

Catalog No. 11392264
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
11-392-264 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-392-264 Supplier Velp™ Scientific Supplier No. F30210125
Add to Cart
Add to Cart

Entry-level distillation for Kjeldahl protein determination, non-protein nitrogen (NPN), volatiles and more. Simple and reliable solution with unique features.

  • Most suitable distiller for laboratories looking for a simple but efficient instrument for Kjeldahl nitrogen TKN, proteins, ammoniacal nitrogen, nitric nitrogen (Devarda), TVBN, sulfites, phenols, volatile acids, cyanides, and alcohol content
  • Programmable sodium hydroxide addition
  • High-precision pump ensuring constant accurate dosing of the reagent
  • Intuitive interface for easy analysis settings
  • Unique VELP™ patented titanium condenser designed for efficient thermal exchange and low water consumption
  • Distillate temperature is always kept below Kjeldahl threshold value (35°C), without nitrogen loss
  • Reduced water consumption
  • High resistance material with prolonged lifespan
  • VELP™ Technopolymer Splash Head is designed for high throughput providing state-of-the-art durability and resistance

Specifications

Voltage 115 V
Type Distillation Unit
Frequency 50/60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.