Learn More
Eurofins Discoverx UDP, 10 mg. (92-1178)
Catalog No. 501999706
Encompass
Uridine 5'-(trihydrogen diphosphate) is a uracil nucleoside diphosphate that is an important agent in glycogenesis in liver and muscles. UDP is a potent agonist of the P2Y purinoceptor. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9706 Supplier Eurofins Discoverx Supplier No. 921178
Uridine 5'-(trihydrogen diphosphate) is a uracil nucleoside diphosphate that is an important agent in glycogenesis in liver and muscles. UDP is a potent agonist of the P2Y purinoceptor. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.