Learn More
Cayman Chemical UDP-a-D-GlucosesodIum salt 1g
SDP
Supplier:  Cayman Chemical 156021 G
Encompass_Preferred
An endogenous nucleotide sugar that binds the P2Y14 receptor (EC50 = 0.35 μM) as well as GPR17; induces oligodendrocyte differentiation at a maximal concentration of 100 μM
Catalog No. 50-296-9930