Learn More
Active Motif UDP-Azide-Glucose
SDP
Catalog No. NC1109893
Encompass
UDP-Azide-Glucose is used in conjunction with Beta-glucosyltransferase enzyme (Catalog No. 55012) to detect and capture DNA fragments containing 5-hydroxymethylcytosine (5-hmC) DNA methylation for analysis. For more complete information, please visit the Active Motif website.
Catalog No. NC1109893 Supplier Active Motif Supplier No. 55020
UDP-Azide-Glucose is used in conjunction with Beta-glucosyltransferase enzyme (Catalog No. 55012) to detect and capture DNA fragments containing 5-hydroxymethylcytosine (5-hmC) DNA methylation for analysis. For more complete information, please visit the Active Motif website.