Learn More
Medchemexpress LLC UDP-α-D-Galactose-13C disodium | 137868-52-1 | 98.0% | 611.26 | 1 MG
SDP
Catalog No. 5000435664
Encompass_Preferred
UDP-α-D-Galactose-13C (disodium) is a 13C labeled UDP-α-D-Galactose disodium salt for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-35664 Supplier Medchemexpress LLC Supplier No. HY114364S1MG
UDP-α-D-Galactose-13C (disodium) is a 13C labeled UDP-α-D-Galactose disodium salt for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs