Learn More
Medchemexpress LLC UDP-D-GalNAlk disodium | 2370947-52-5 | 98.9% | 689.37 | C20H27N3Na2O17P2 | 10 MG
SDP
Catalog No. 5000289301
Encompass_Preferred
UDP-D-GalNAlk disodium is an alkyne-functionalized UDP-galactose derivative (disodium salt) intended for biochemical research. It functions as a substrate for enzymatic glycosylation and enables bioorthogonal labeling through terminal alkyne click chemistry. The compound is supplied as a solid with high reported purity and should be stored sealed and protected from moisture according to supplier recommendations.
  • Alkyne-modified UDP-sugar for bioorthogonal labeling.
  • Serves as substrate for galactosyltransferase and glycosylation assays.
  • High purity suitable for research applications.
  • Available in small milligram quantities for lab use.
  • Stable when stored sealed and kept away from moisture as directed.
Catalog No. 50-002-89301 Supplier Medchemexpress LLC Supplier No. HY164903A10MG
UDP-D-GalNAlk disodium is an alkyne-functionalized UDP-galactose derivative (disodium salt) intended for biochemical research. It functions as a substrate for enzymatic glycosylation and enables bioorthogonal labeling through terminal alkyne click chemistry. The compound is supplied as a solid with high reported purity and should be stored sealed and protected from moisture according to supplier recommendations.
  • Alkyne-modified UDP-sugar for bioorthogonal labeling.
  • Serves as substrate for galactosyltransferase and glycosylation assays.
  • High purity suitable for research applications.
  • Available in small milligram quantities for lab use.
  • Stable when stored sealed and kept away from moisture as directed.