Learn More
Selleck Chemical LLC Uridine-5-diphosphate disodium salt S3368-1g
SDP
Catalog No. 503109535
Encompass_Preferred
Uridine-5 -diphosphate (UDP) disodium salt is a potent selective and natural agonist of P2Y6 receptor with EC50 300 nM and pEC50 of 6 52 respectively Uridine-5 -diphosphate (UDP) disodium salt is also an antagonist of P2Y14 receptor with pEC50 of 7 28
Catalog No. 50-310-9535 Supplier Selleck Chemical LLC Supplier No. S33681G
Uridine-5 -diphosphate (UDP) disodium salt is a potent selective and natural agonist of P2Y6 receptor with EC50 300 nM and pEC50 of 6 52 respectively Uridine-5 -diphosphate (UDP) disodium salt is also an antagonist of P2Y14 receptor with pEC50 of 7 28