Learn More
Apexbio Technology LLC UDP disodium salt(Synonyms: Uridine 5'-diphosphate disodium salt, Uridine diphosphate disodium salt, UDP-Na2), 50mg, CAS: 27821-45-0.
SDP
Catalog No. 5000061198
Encompass_Preferred
UDP disodium salt (CAS 27821-45-0) is a selective agonist targeting the P2Y6 receptor exhibiting potent stimulatory activity with an EC50 value of approximately 300 nM and a reported pEC50 of 6 52 for the human P2Y6 receptor Endogenously UDP serves as a metabolite involved in catalyzing glucuronidation reactions facilitating the conjugation of glucuronic acid to diverse substrates Due to these biological properties UDP disodium salt finds utility in researching disease-related mechanisms and RNA biosynthesis pathways
Catalog No. 50-000-61198 Supplier Apexbio Technology LLC Supplier No. B727850
UDP disodium salt (CAS 27821-45-0) is a selective agonist targeting the P2Y6 receptor exhibiting potent stimulatory activity with an EC50 value of approximately 300 nM and a reported pEC50 of 6 52 for the human P2Y6 receptor Endogenously UDP serves as a metabolite involved in catalyzing glucuronidation reactions facilitating the conjugation of glucuronic acid to diverse substrates Due to these biological properties UDP disodium salt finds utility in researching disease-related mechanisms and RNA biosynthesis pathways