Learn More
American Radiolabeled Chemicals Inc UDP GAL-1-14C 50UC
SDP
Catalog No. 50901278
Encompass_Preferred
Uridine 5'-diphosphate galactose-1-14C 50uCi in ethanol:water (7:3), 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 566.3, Shipped in Dry Ice
Catalog No. 50-901-278 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 347250UC
Add to Cart
Add to Cart
Request a Quote
Uridine 5'-diphosphate galactose-1-14C 50uCi in ethanol:water (7:3), 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 566.3, Shipped in Dry Ice