Learn More
AdipoGen UDP-galactose
SDP
Catalog No. 502559927
Encompass_Preferred
Chemical. CAS 137868-52-1. Formula C15H22N2Na2O17P2. MW 610.27. Uridine 5-diphospho-alpha-D-galactose UDP-Gal is a nucleotide sugar. UDP-Gal is a donor substrate for galactosyltransferases used in the biosynthesis of galactose-containing oligosaccharides. UDP-Gal has been used as a component in galactosyltransferase labeling buffer to prelabel terminal N-acetylglucosamine GlcNAc on immunoglobulin chains. It has also been used as a component in Hyp O-galactosyltransferase HPGT assay. Decreased red blood cell levels of UDP-Gal are found in patients with maple syrup urine disease or phenylketonuria.
Catalog No. 50-255-9927 Supplier AdipoGen Supplier No. CDXU0009M005
Chemical. CAS 137868-52-1. Formula C15H22N2Na2O17P2. MW 610.27. Uridine 5-diphospho-alpha-D-galactose UDP-Gal is a nucleotide sugar. UDP-Gal is a donor substrate for galactosyltransferases used in the biosynthesis of galactose-containing oligosaccharides. UDP-Gal has been used as a component in galactosyltransferase labeling buffer to prelabel terminal N-acetylglucosamine GlcNAc on immunoglobulin chains. It has also been used as a component in Hyp O-galactosyltransferase HPGT assay. Decreased red blood cell levels of UDP-Gal are found in patients with maple syrup urine disease or phenylketonuria.