Learn More
Medchemexpress LLC UDP-Galactose disodi 100mg | 137868-52-1 | 100 MG
SDP
Catalog No. 5000307753
Encompass_Preferred
Udp-galactose disodium is a nucleotide sugar used as a donor substrate for galactosyltransferases and an endogenous agonist of the P2Y14 receptor. It is commonly used in glycobiology and enzymology research.
  • Serves as a donor substrate for beta-1,4-galactosyltransferases.
  • Acts as a P2Y14 receptor agonist (hP2Y14 EC50 ~0.67 μM).
  • High purity suitable for biochemical assays.
  • Available in multiple sizes, including 100 mg.
  • Stable when stored sealed at 4°C; in solution, store at -80°C for long-term storage.
Catalog No. 50-003-07753 Supplier Medchemexpress LLC Supplier No. HY114364100MG
Udp-galactose disodium is a nucleotide sugar used as a donor substrate for galactosyltransferases and an endogenous agonist of the P2Y14 receptor. It is commonly used in glycobiology and enzymology research.
  • Serves as a donor substrate for beta-1,4-galactosyltransferases.
  • Acts as a P2Y14 receptor agonist (hP2Y14 EC50 ~0.67 μM).
  • High purity suitable for biochemical assays.
  • Available in multiple sizes, including 100 mg.
  • Stable when stored sealed at 4°C; in solution, store at -80°C for long-term storage.