Learn More
Medchemexpress LLC UDP-Galactose disodi 10mg | 137868-52-1 | 10 MG
SDP
Catalog No. 5000307570
Encompass_Preferred
UDP-galactose disodium is the disodium salt of uridine diphosphate galactose, a nucleotide sugar used as a donor substrate for galactosyltransferases and as a P2Y14 receptor agonist. It is supplied as a white to off-white solid for biochemical and enzymatic assays.
  • High purity suitable for enzymatic assays.
  • White to off-white solid form for easy handling.
  • Molecular weight 610.27 and formula C15H22N2Na2O17P2.
  • Stable when stored sealed away from moisture and light; in solution, store at -80°C for long-term.
  • Used as a donor substrate for galactosyltransferases and in receptor agonist studies.
Catalog No. 50-003-07570 Supplier Medchemexpress LLC Supplier No. HY11436410MG
UDP-galactose disodium is the disodium salt of uridine diphosphate galactose, a nucleotide sugar used as a donor substrate for galactosyltransferases and as a P2Y14 receptor agonist. It is supplied as a white to off-white solid for biochemical and enzymatic assays.
  • High purity suitable for enzymatic assays.
  • White to off-white solid form for easy handling.
  • Molecular weight 610.27 and formula C15H22N2Na2O17P2.
  • Stable when stored sealed away from moisture and light; in solution, store at -80°C for long-term.
  • Used as a donor substrate for galactosyltransferases and in receptor agonist studies.