Learn More
Medchemexpress LLC UDP-Galactose disodi 5mg | 137868-52-1 | 5 MG
SDP
Catalog No. 5000226922
Encompass_Preferred
UDP-Galactose disodium is the disodium salt of uridine diphosphate galactose, a nucleotide sugar used as a galactose donor in enzymatic glycosylation and as a P2Y14 receptor agonist for biochemical and cell-signaling research.
  • Donor substrate for galactosyltransferases and glycoconjugate biosynthesis.
  • Agonist of the P2Y14 receptor (EC50 0.67 μM for hP2Y14).
  • High purity solid suitable for biochemical assays (99.4%).
  • White to off-white solid, supplied as a five mg quantity.
  • Storage: 4°C sealed; in solvent store at -80°C for up to six months or -20°C for up to one month.
Catalog No. 50-002-26922 Supplier Medchemexpress LLC Supplier No. HY1143645MG
UDP-Galactose disodium is the disodium salt of uridine diphosphate galactose, a nucleotide sugar used as a galactose donor in enzymatic glycosylation and as a P2Y14 receptor agonist for biochemical and cell-signaling research.
  • Donor substrate for galactosyltransferases and glycoconjugate biosynthesis.
  • Agonist of the P2Y14 receptor (EC50 0.67 μM for hP2Y14).
  • High purity solid suitable for biochemical assays (99.4%).
  • White to off-white solid, supplied as a five mg quantity.
  • Storage: 4°C sealed; in solvent store at -80°C for up to six months or -20°C for up to one month.