Learn More
Medchemexpress LLC UDP-GalNAc disodium 10mg | 108320-87-2 | 10 MG
SDP
Catalog No. 5000307754
Encompass_Preferred
UDP-N-acetyl-D-galactosamine disodium is a sugar nucleotide substrate commonly used in glycobiology and enzymology research.
  • High purity solid suitable for biochemical assays.
  • Molecular weight 651.32 g/mol and formula C17H25N3Na2O17P2.
  • Available in small research quantities, including 10 mg pack sizes.
  • Store sealed and away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Used as a substrate for glycosyltransferases and related enzymatic studies.
Catalog No. 50-003-07754 Supplier Medchemexpress LLC Supplier No. HY11436510MG
UDP-N-acetyl-D-galactosamine disodium is a sugar nucleotide substrate commonly used in glycobiology and enzymology research.
  • High purity solid suitable for biochemical assays.
  • Molecular weight 651.32 g/mol and formula C17H25N3Na2O17P2.
  • Available in small research quantities, including 10 mg pack sizes.
  • Store sealed and away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Used as a substrate for glycosyltransferases and related enzymatic studies.