Learn More
Cayman Chemical UDP-GalNAc (sodium salt), Purity: ≥90%, CAS: 108320-87-2, 10 mg
SDP
Catalog No. 502912317
Encompass_Preferred
A nucleotide sugar; used by glycosyltransferases to glycosylate various substrates
Catalog No. 50-291-2317 Supplier Cayman Chemical Supplier No. 3650610 MG
A nucleotide sugar; used by glycosyltransferases to glycosylate various substrates