Learn More
Cayman Chemical UDP-GalNAc (sodium salt), Purity: ≥90%, CAS: 108320-87-2, 1 mg
SDP
Catalog No. 502914052
Encompass_Preferred
A nucleotide sugar; used by glycosyltransferases to glycosylate various substrates
Catalog No. 50-291-4052 Supplier Cayman Chemical Supplier No. 365061 MG
A nucleotide sugar; used by glycosyltransferases to glycosylate various substrates