Learn More
Medchemexpress LLC UDP-GlcNAc-13C disodium | 91183-98-1 | 97.0% | 652.31 | 1 MG
SDP
Catalog No. 5000438085
Encompass_Preferred
UDP-GlcNAc-13C (disodium) is the 13C labeled UDP-GlcNAc Disodium Salt, a donor substrate of O-GlcNAc transferase (O-GlcNAc). It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-38085 Supplier Medchemexpress LLC Supplier No. HY112174S1MG
UDP-GlcNAc-13C (disodium) is the 13C labeled UDP-GlcNAc Disodium Salt, a donor substrate of O-GlcNAc transferase (O-GlcNAc). It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.