Learn More
Medchemexpress LLC UDP-GlcNAc disodium | 91183-98-1 | 99.9% | 651.32 | 10 MG
SDP
Catalog No. 5000438373
Encompass_Preferred
UDP-GlcNAc Disodium Salt is a donor substrate of O-GlcNAc transferase (OGT). OGT is a nucleocytoplasmic glycosyltransferase that modifies thousands of proteins by adding a unique N-acetylglucosamine residue onto acceptor substrates mainly confined within the cytosol and nucleus.
  • Donor substrate of O-GlcNAc transferase (OGT).
  • For research use only.
Catalog No. 50-004-38373 Supplier Medchemexpress LLC Supplier No. HY11217410MG
UDP-GlcNAc Disodium Salt is a donor substrate of O-GlcNAc transferase (OGT). OGT is a nucleocytoplasmic glycosyltransferase that modifies thousands of proteins by adding a unique N-acetylglucosamine residue onto acceptor substrates mainly confined within the cytosol and nucleus.
  • Donor substrate of O-GlcNAc transferase (OGT).
  • For research use only.