Learn More
Medchemexpress LLC UDP-GlcNAc disodium | 91183-98-1 | 99.9% | 651.32 | 100 MG
SDP
Catalog No. 5000438273
Encompass_Preferred
UDP-GlcNAc Disodium Salt is a donor substrate of O-GlcNAc transferase (OGT). OGT is a nucleocytoplasmic glycosyltransferase that modifies thousands of proteins by adding a unique N-acetylglucosamine residue onto acceptor substrates, mainly confined within the cytosol and nucleus.
  • Donor substrate of O-GlcNAc transferase (OGT).
  • For research use only.
Catalog No. 50-004-38273 Supplier Medchemexpress LLC Supplier No. HY112174100MG
UDP-GlcNAc Disodium Salt is a donor substrate of O-GlcNAc transferase (OGT). OGT is a nucleocytoplasmic glycosyltransferase that modifies thousands of proteins by adding a unique N-acetylglucosamine residue onto acceptor substrates, mainly confined within the cytosol and nucleus.
  • Donor substrate of O-GlcNAc transferase (OGT).
  • For research use only.