Learn More
TARGETMOL CHEMICALS INC UDP-GlcNAc Disodium Salt 10MG
SDP
Catalog No. 502878343
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. UDP-GlcNAc Disodium Salt (UDP-alpha-D-N-Acetylglucosamine Disodium Salt) is a nucleotide sugar synthesized from glucose through the hexosamine biosynthetic pathway (HBP). Purity 99.2%
Catalog No. 50-287-8343 Supplier TARGETMOL CHEMICALS INC Supplier No. T1959610MG
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. UDP-GlcNAc Disodium Salt (UDP-alpha-D-N-Acetylglucosamine Disodium Salt) is a nucleotide sugar synthesized from glucose through the hexosamine biosynthetic pathway (HBP). Purity 99.2%