Learn More
Medchemexpress LLC UDP-GlcNAz disodium | 1611490-64-2 | 99.7% | C17H24N6Na2O17P2 | 100 MG
SDP
Catalog No. 5000353965
Encompass_Preferred
UDP-GlcNAz disodium is an analogue of UDP-GlcNAc disodium. It functions as a donor substrate for various N-acetylgalactosaminyltransferases and is a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Functions as a sugar donor
  • Serves as a substrate for OGTase and O-GlcNAcase
  • Capable of glycosylating p62
  • Glycosidic bond can be cleaved by O-GlcNAcase catalysis
  • Useful in research concerning protein glycosylation
  • Can be incorporated into nuclear proteins in Jurkat cells
Catalog No. 50-003-53965 Supplier Medchemexpress LLC Supplier No. HY145934A100MG
UDP-GlcNAz disodium is an analogue of UDP-GlcNAc disodium. It functions as a donor substrate for various N-acetylgalactosaminyltransferases and is a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Functions as a sugar donor
  • Serves as a substrate for OGTase and O-GlcNAcase
  • Capable of glycosylating p62
  • Glycosidic bond can be cleaved by O-GlcNAcase catalysis
  • Useful in research concerning protein glycosylation
  • Can be incorporated into nuclear proteins in Jurkat cells