Learn More
Medchemexpress LLC UDP-GlcNAz disodium | 1611490-64-2 | 99.7% | C17H24N6Na2O17P2 | 25 MG
SDP
Catalog No. 5000353070
Encompass_Preferred
UDP-GlcNAz disodium is an analogue of UDP-GlcNAc disodium. It is a click chemistry reagent, capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. It also acts as a sugar donor and a substrate for OGTase and O-GlcNAcase, valuable in protein glycosylation research.
  • Analogue of UDP-GlcNAc disodium
  • Functions as a click chemistry reagent
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Acts as a sugar donor
  • Serves as a substrate for OGTase and O-GlcNAcase
  • Can glycosylate p62
  • Glycosidic bond can be cleaved by O-GlcNAcase through catalysis
  • Incorporated into nuclear proteins in Jurkat cells in *in vitro* studies
Catalog No. 50-003-53070 Supplier Medchemexpress LLC Supplier No. 25MGHY145934A
UDP-GlcNAz disodium is an analogue of UDP-GlcNAc disodium. It is a click chemistry reagent, capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. It also acts as a sugar donor and a substrate for OGTase and O-GlcNAcase, valuable in protein glycosylation research.
  • Analogue of UDP-GlcNAc disodium
  • Functions as a click chemistry reagent
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Acts as a sugar donor
  • Serves as a substrate for OGTase and O-GlcNAcase
  • Can glycosylate p62
  • Glycosidic bond can be cleaved by O-GlcNAcase through catalysis
  • Incorporated into nuclear proteins in Jurkat cells in *in vitro* studies