Learn More
Medchemexpress LLC UDP-GlcNAz disodium (UDP-N-azidoacetylgalactosamine disodium) | 1611490-64-2 | 99.7% | C17H24N6Na2O17P2 | 10MG
SDP
Catalog No. 5000261563
Encompass_Preferred
UDP-GlcNAz disodium is an azide-modified uridine diphosphate sugar (disodium salt) used as a donor substrate for glycosyltransferases in biochemical and enzymology studies. It enables incorporation of an azido group into glycans for downstream bioorthogonal labeling and analysis.
  • Azide functional group enables bioorthogonal click chemistry labeling.
  • Serves as a substrate for O-GlcNAc transferase and other glycosyltransferases.
  • High purity suitable for analytical and biochemical assays.
  • Disodium salt form improves aqueous solubility and stability.
  • Available in small-scale lab pack sizes for research applications.
Catalog No. 50-002-61563 Supplier Medchemexpress LLC Supplier No. HY145934A10MG
UDP-GlcNAz disodium is an azide-modified uridine diphosphate sugar (disodium salt) used as a donor substrate for glycosyltransferases in biochemical and enzymology studies. It enables incorporation of an azido group into glycans for downstream bioorthogonal labeling and analysis.
  • Azide functional group enables bioorthogonal click chemistry labeling.
  • Serves as a substrate for O-GlcNAc transferase and other glycosyltransferases.
  • High purity suitable for analytical and biochemical assays.
  • Disodium salt form improves aqueous solubility and stability.
  • Available in small-scale lab pack sizes for research applications.