Learn More
Medchemexpress LLC Udp-glucosamine disodium | 1355005-51-4 | MFCD06809590 | 99.4% | 609.28 g·mol⁻¹ | C15H23N3Na2O16P2 | 5 MG
SDP
Catalog No. 5000223297
Encompass_Preferred
UDP-glucosamine disodium is the disodium salt of UDP-glucosamine (UDP-GlcNAc), a nucleotide sugar used as a substrate for O-GlcNAc transferase in biochemical and enzymatic assays. It is supplied as a white to off-white solid for research applications, including studies of protein O-GlcNAcylation and glycosylation pathways.
  • High purity (99.43%) suitable for enzymatic and biochemical assays.
  • Supplied as a dry solid for convenient storage and handling.
  • Available in small milligram package sizes for research-scale use.
  • Molecular weight 609.28 g·mol⁻¹; formula C15H23N3Na2O16P2.
Catalog No. 50-002-23297 Supplier Medchemexpress LLC Supplier No. HY1397935MG
UDP-glucosamine disodium is the disodium salt of UDP-glucosamine (UDP-GlcNAc), a nucleotide sugar used as a substrate for O-GlcNAc transferase in biochemical and enzymatic assays. It is supplied as a white to off-white solid for research applications, including studies of protein O-GlcNAcylation and glycosylation pathways.
  • High purity (99.43%) suitable for enzymatic and biochemical assays.
  • Supplied as a dry solid for convenient storage and handling.
  • Available in small milligram package sizes for research-scale use.
  • Molecular weight 609.28 g·mol⁻¹; formula C15H23N3Na2O16P2.