Learn More
Medchemexpress LLC UDP-glucose dehydrogenase | 9028-26-6 | 10 UG
SDP
Catalog No. 5000353436
Encompass_Preferred
UDP-glucose dehydrogenase is a cytosolic enzyme that catalyzes the oxidation of UDP-glucose to UDP-glucuronate. It plays a critical role in the synthesis of glycosaminoglycans and proteoglycans.
  • Catalyzes the oxidation of UDP-glucose to UDP-glucuronate.
  • Plays a critical role in synthesis of glycosaminoglycans and proteoglycans.
  • For research use only.
Catalog No. 50-003-53436 Supplier Medchemexpress LLC Supplier No. HY14185110UG
UDP-glucose dehydrogenase is a cytosolic enzyme that catalyzes the oxidation of UDP-glucose to UDP-glucuronate. It plays a critical role in the synthesis of glycosaminoglycans and proteoglycans.
  • Catalyzes the oxidation of UDP-glucose to UDP-glucuronate.
  • Plays a critical role in synthesis of glycosaminoglycans and proteoglycans.
  • For research use only.