Learn More
Medchemexpress LLC UDP-glucose dehydrogenase | 9028-26-6 | 25UG
SDP
Catalog No. 5000353004
Encompass_Preferred
UDP-glucose dehydrogenase is a cytosolic enzyme that catalyzes the oxidation of UDP-glucose to UDP-glucuronate. It plays a critical role in the synthesis of glycosaminoglycans and proteoglycans.
  • For research use only
Catalog No. 50-003-53004 Supplier Medchemexpress LLC Supplier No. 25UGHY141851
UDP-glucose dehydrogenase is a cytosolic enzyme that catalyzes the oxidation of UDP-glucose to UDP-glucuronate. It plays a critical role in the synthesis of glycosaminoglycans and proteoglycans.
  • For research use only