Learn More
Medchemexpress LLC UDP-glucose dehydrogenase | 9028-26-6 | 50 UG
SDP
Catalog No. 5000353016
Encompass_Preferred
UDP-glucose dehydrogenase is a cytosolic enzyme that catalyzes the oxidation of UDP-glucose to UDP-glucuronate. It plays a critical role in the synthesis of glycosaminoglycans and proteoglycans. This product is for research use only and not intended for human use.
  • Enzyme
  • Biochemical detection
  • Recombinant enzyme
  • Metabolic disease research
  • Metabolic enzyme/protease
  • Endogenous metabolite
  • Inhibitor
Catalog No. 50-003-53016 Supplier Medchemexpress LLC Supplier No. HY14185150UG
UDP-glucose dehydrogenase is a cytosolic enzyme that catalyzes the oxidation of UDP-glucose to UDP-glucuronate. It plays a critical role in the synthesis of glycosaminoglycans and proteoglycans. This product is for research use only and not intended for human use.
  • Enzyme
  • Biochemical detection
  • Recombinant enzyme
  • Metabolic disease research
  • Metabolic enzyme/protease
  • Endogenous metabolite
  • Inhibitor