Learn More
Abcam UDP-glucose, Highly P2Y14 agonist, 100MG
Catalog No. 502584612
Encompass_Preferred
MW 610.27 Da. Highly potent, endogenous P2Y14 agonist (EC50 = 82 nM). Involved in modulation of gastric function. UDP also available (ab120383).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4612 Supplier Abcam Supplier No. AB120384100MG
MW 610.27 Da. Highly potent, endogenous P2Y14 agonist (EC50 = 82 nM). Involved in modulation of gastric function. UDP also available (ab120383).
The product is subject to the following: Abcam Restricted Use Statement