Learn More
Abcam UDP-glucose, Highly P2Y14 agonist, 500MG
Catalog No. 502584610
Encompass_Preferred
MW 610.27 Da. Highly potent, endogenous P2Y14 agonist (EC50 = 82 nM). Involved in modulation of gastric function. UDP also available (ab120383).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4610 Supplier Abcam Supplier No. AB120384500MG
MW 610.27 Da. Highly potent, endogenous P2Y14 agonist (EC50 = 82 nM). Involved in modulation of gastric function. UDP also available (ab120383).
The product is subject to the following: Abcam Restricted Use Statement