Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UDP-glucuronic acid trisodium | 63700-19-6 | 100 MG
SDP

Catalog No. 5000418252
Encompass_Preferred

UDP-glucuronic acid trisodium (Uridine 5'-diphosphoglucuronic acid trisodium salt) is a crucial precursor for vital glycoconjugates found across biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids. It is intended for research use only.

  • Promotes migration of liver cancer cells (Huh7, SK-Hep1, SNU449) by inducing EMT process.
  • Molecular Formula: C15H19N2Na3O18P2.
  • Molecular Weight: 646.23.
  • Target: Drug intermediate.
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (20 mg/mL).
  • Store solid at -20°C; store in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-18252 Supplier Medchemexpress LLC Supplier No. HYN7033100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UDP-glucuronic acid trisodium (Uridine 5'-diphosphoglucuronic acid trisodium salt) is a crucial precursor for vital glycoconjugates found across biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids. It is intended for research use only.

  • Promotes migration of liver cancer cells (Huh7, SK-Hep1, SNU449) by inducing EMT process.
  • Molecular Formula: C15H19N2Na3O18P2.
  • Molecular Weight: 646.23.
  • Target: Drug intermediate.
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (20 mg/mL).
  • Store solid at -20°C; store in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.