Learn More
Medchemexpress LLC UDP-glucuronic acid trisodium | 63700-19-6 | 25 MG
SDP
Catalog No. 5000421823
Encompass_Preferred
UDP-glucuronic acid trisodium (Uridine-5'-diphosphoglucuronic acid trisodium salt) is a critical precursor for essential glycoconjugates across biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids.
  • Promotes the migration of liver cancer cells (Huh7, SK-Hep1, and SNU449) at concentrations of 0.5-1 mM over 24 hours.
  • Induces the EMT process.
  • Decreased E-cadherin levels and increased vimentin levels in Huh7 and SNU449 cells at 0.5 mM over 24 hours.
Catalog No. 50-004-21823 Supplier Medchemexpress LLC Supplier No. 25MGHYN7033
UDP-glucuronic acid trisodium (Uridine-5'-diphosphoglucuronic acid trisodium salt) is a critical precursor for essential glycoconjugates across biological kingdoms, including mammalian glycosaminoglycans, plant cell wall polysaccharides, and bacterial capsule glycoglycerolipids.
  • Promotes the migration of liver cancer cells (Huh7, SK-Hep1, and SNU449) at concentrations of 0.5-1 mM over 24 hours.
  • Induces the EMT process.
  • Decreased E-cadherin levels and increased vimentin levels in Huh7 and SNU449 cells at 0.5 mM over 24 hours.